Assay Details
Target Gene Details
Entrez Gene ID: | 22209 |
Gene Name: | ubiquitin-conjugating enzyme E2A |
Gene Aliases: |
HHR6A, HR6A, Mhr6a |
Location: |
Chr.X:36874366-36884222 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2a | NM_019668.3 | NP_062642.1 | ||
| AF089812.1 | AAC64563.1 | |||
| AF383148.1 | AAK62984.1 | |||
| AK133723.1 | BAE21800.1 | |||
| AK150417.1 | BAE29541.1 | |||
| AK153327.1 | BAE31906.1 | |||
| AK160527.1 | BAE35845.1 | |||
| AK160538.1 | BAE35855.1 | |||
| AK160545.1 | BAE35862.1 | |||
| AK163966.1 | BAE37558.1 | |||
| AK164074.1 | BAE37614.1 | |||
| BC026053.1 | AAH26053.1 |

Genomic Map