Assay Details
Target Gene Details
Entrez Gene ID: | 208968 |
Gene Name: | zinc finger protein 280C |
Gene Aliases: |
BC028839, Suhw3, Zfp633, Znf280c, mKIAA1584 |
Location: |
Chr.X:48541626-48594646 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp280c | NM_001166648.1 | NP_001160120.1 | ||
| NM_001166649.1 | NP_001160121.1 | |||
| NM_001166650.1 | NP_001160122.1 | |||
| NM_153532.3 | NP_705760.2 | |||
| AK129399.1 | BAC98209.1 | |||
| AK141491.1 | BAE24700.1 | |||
| BC028839.1 | AAH28839.1 | |||
| BC051397.1 | AAH51397.1 | |||
| BC063775.1 | AAH63775.1 | |||
| BY016949.1 |
More Information
Set Membership: |
|

Genomic Map