Assay Details
Target Gene Details
Entrez Gene ID: | 14660 |
Gene Name: | glutaminase |
Gene Aliases: |
6330442B14, AI314027, B230365M23Rik |
Location: |
Chr.1:52163449-52233232 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gls | NM_001081081.2 | NP_001074550.1 | ||
| NM_001113383.1 | 15 | 2740 | NP_001106854.1 | |
| AK046297.1 | 10 | 1662 | ||
| AK048072.1 | 13 | 1918 | ||
| AK049883.1 | ||||
| AK088231.1 | 1 | 648 | ||
| AK173039.1 | 15 | 2740 | BAD32317.1 |
More Information
Set Membership: |
|

Genomic Map