Assay Details
Target Gene Details
Entrez Gene ID: | 227399 |
Gene Name: | diphosphoinositol pentakisphosphate kinase 2 |
Gene Aliases: |
AW555814, D330021B20, Hisppd1, Vip2, mKIAA0433 |
Location: |
Chr.1:97706043-97770250 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppip5k2 | NM_173760.5 | 28 | 4380 | NP_776121.4 |
| AK052286.1 | 7 | 1177 | BAC34917.1 | |
| AK087712.1 | 28 | 4312 | ||
| AK129140.1 | 28 | 4316 | BAC97950.1 | |
| AK137392.1 | 3 | 2872 | ||
| BC046411.1 | 18 | 2693 | AAH46411.1 | |
| BC053396.1 | 27 | 3882 | AAH53396.1 |
More Information
Set Membership: |
|

Genomic Map