Assay Details
Target Gene Details
Entrez Gene ID: | 226422 |
Gene Name: | RAB29, member RAS oncogene family |
Gene Aliases: |
Rab7l1 |
Location: |
Chr.1:131867210-131872887 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab29 | NM_144875.2 | 6 | 1048 | NP_659124.2 |
| AK079752.1 | 5 | 902 | ||
| AK185980.1 | 1 | 273 | ||
| BC016133.1 | 6 | 1033 | AAH16133.1 | |
| BC029056.1 | 5 | 822 | AAH29056.1 | |
| CK796080.1 | 1 | 167 |

Genomic Map