Assay Details
Target Gene Details
Entrez Gene ID: | 64143 |
Gene Name: | v-ral simian leukemia viral oncogene homolog B (ras related) |
Gene Aliases: |
5730472O18Rik |
Location: |
Chr.1:119470305-119504782 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralb | NM_022327.5 | 5 | 1012 | NP_071722.1 |
| AK017698.1 | 5 | 858 | BAB30881.1 | |
| AK033193.1 | 5 | 867 | BAC28193.1 | |
| AK159993.1 | 5 | 957 | BAE35543.1 | |
| BC006907.1 | 5 | 856 | AAH06907.1 |
More Information
Set Membership: |
|

Genomic Map