Assay Details
Target Gene Details
Entrez Gene ID: | 13710 |
Gene Name: | E74-like factor 3 |
Gene Aliases: |
ESE-1, ESX, jen |
Location: |
Chr.1:135253574-135258565 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elf3 | NM_001163131.1 | 9 | 1471 | NP_001156603.1 |
| NM_007921.3 | 9 | 1411 | NP_031947.1 | |
| AF016294.1 | 9 | 1377 | AAB96585.1 | |
| AK143135.1 | 9 | 1461 | BAE25282.1 | |
| BC138692.1 | 9 | 1375 | ||
| BC145380.1 | 9 | 1435 |
More Information
Set Membership: |
|

Genomic Map