Assay Details
Target Gene Details
Entrez Gene ID: | 13714 |
Gene Name: | ELK4, member of ETS oncogene family |
Gene Aliases: |
2310011G17Rik, A130026I01Rik, BB162516, SAP-1, Sap1 |
Location: |
Chr.1:132007234-132032612 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk4 | NM_007923.2 | 1 | 281 | NP_031949.2 |
| AK156537.1 | 1 | 279 | ||
| BC144844.1 | 1 | 31 | ||
| BC145795.1 | 1 | 31 | ||
| Z36885.1 | CAA85358.1 |
More Information
Set Membership: |
|

Genomic Map