Assay Details
Target Gene Details
Entrez Gene ID: | 13714 |
Gene Name: | ELK4, member of ETS oncogene family |
Gene Aliases: |
2310011G17Rik, A130026I01Rik, BB162516, SAP-1, Sap1 |
Location: |
Chr.1:132007234-132032612 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk4 | NM_007923.2 | 3 | 1271 | NP_031949.2 |
| AK054075.1 | BAC35647.2 | |||
| AK156537.1 | 3 | 1269 | ||
| BC004798.1 | 1 | 516 | AAH04798.1 | |
| BC144844.1 | 3 | 1021 | ||
| BC145795.1 | 3 | 1021 | ||
| Z36885.1 | 4 | 1216 | CAA85358.1 |
More Information
Set Membership: |
|

Genomic Map