Assay Details
Target Gene Details
Entrez Gene ID: | 215690 |
Gene Name: | neuron navigator 1 |
Gene Aliases: |
9530089B19, 9930003A20Rik, C230080M11Rik, Pomfil3, Unc53h1, mKIAA1151, steerin-1 |
Location: |
Chr.1:135434580-135688116 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nav1 | NM_173437.2 | 30 | 9166 | NP_775613.2 |
| AK137320.1 | 1 | 751 |

Genomic Map