Assay Details
Target Gene Details
Entrez Gene ID: | 66352 |
Gene Name: | basic leucine zipper nuclear factor 1 |
Gene Aliases: |
1700030G05Rik, Blzf1l, Golgin-45, Jem-1 |
Location: |
Chr.1:164289800-164307484 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blzf1 | NM_001160208.1 | 8 | 2186 | NP_001153680.1 |
| NM_001160209.1 | 7 | 2273 | NP_001153681.1 | |
| NM_025505.4 | 9 | 2207 | NP_079781.2 | |
| AK005489.1 | 7 | 2007 | BAB24074.1 | |
| AK029787.1 | 9 | 2208 | BAC26617.1 | |
| AK167493.1 | 7 | 2280 | BAE39571.1 | |
| AK168933.1 | 7 | 2034 | ||
| BC027025.1 | 7 | 2048 | AAH27025.1 |
More Information
Set Membership: |
|

Genomic Map