Assay Details
Target Gene Details
Entrez Gene ID: | 280287 |
Gene Name: | KiSS-1 metastasis-suppressor |
Gene Aliases: |
kisspeptin, metastatin |
Location: |
Chr.1:133321887-133329722 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kiss1 | NM_178260.3 | 2 | 190 | NP_839991.2 |
| AA023297.1 | ||||
| AB162440.1 | 2 | 152 | BAD36756.1 | |
| AB666166.1 | 3 | 350 | ||
| AF472576.1 | 2 | 202 | AAP13356.1 | |
| BC117046.1 | 2 | 187 | ||
| BC119348.1 | 2 | 187 |

Genomic Map