Assay Details
Target Gene Details
Entrez Gene ID: | 17970 |
Gene Name: | neutrophil cytosolic factor 2 |
Gene Aliases: |
NOXA2, Ncf-2, p67phox |
Location: |
Chr.1:152800158-152837169 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncf2 | NM_010877.4 | 15 | 2255 | NP_035007.1 |
| AK036379.1 | 15 | 2256 | BAC29404.1 | |
| AK155324.1 | 15 | 2310 | BAE33191.1 |
More Information
Set Membership: |
|

Genomic Map