Assay Details
Target Gene Details
Entrez Gene ID: | 11539 |
Gene Name: | adenosine A1 receptor |
Gene Aliases: |
A1-AR, A1AR, A1R, AA1R, AI848715, BB176431, Ri |
Location: |
Chr.1:134199215-134235457 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adora1 | NM_001008533.3 | 1 | 267 | NP_001008533.1 |
| NM_001039510.2 | NP_001034599.1 | |||
| NM_001282945.1 | NP_001269874.1 | |||
| NM_001291930.1 | NP_001278859.1 | |||
| AK038774.1 | BAC30128.1 | |||
| AK043954.1 | 1 | 208 | BAC31713.1 | |
| AK134532.1 | ||||
| AU080986.1 | ||||
| BY257408.1 | 1 | 245 | ||
| KC881107.1 |
More Information
Set Membership: |
|

Genomic Map