Assay Details
Target Gene Details
Entrez Gene ID: | 170788 |
Gene Name: | crumbs family member 1, photoreceptor morphogenesis associated |
Gene Aliases: |
7530426H14Rik, A930008G09Rik |
Location: |
Chr.1:139197546-139379118 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crb1 | NM_133239.2 | 6 | 1495 | NP_573502.2 |
| AF406641.1 | 6 | 1460 | AAL65131.1 | |
| AF492496.1 | 5 | 1336 | AAQ06606.1 | |
| AK044345.1 | 2 | 334 | BAC31879.1 | |
| AK044704.1 | 2 | 373 | BAC32041.1 | |
| AY450552.1 | 6 | 1460 | AAR20495.1 |
More Information
Set Membership: |
|

Genomic Map