Assay Details
Target Gene Details
Entrez Gene ID: | 381314 |
Gene Name: | isoleucine-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
2010002H18Rik, C79125 |
Location: |
Chr.1:185286662-185329419 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iars2 | NM_198653.2 | 23 | 3463 | NP_941055.1 |
| AK036182.1 | 23 | 3431 | BAC29337.1 | |
| AK043590.1 | 23 | 3465 | ||
| AK048653.1 | 23 | 3464 | BAC33410.1 | |
| AK050358.1 | 23 | 3446 | BAC34208.1 | |
| AK078183.1 | ||||
| AK085714.1 | ||||
| AK088965.1 | ||||
| AK166832.1 | BAE39055.1 | |||
| BC027432.1 | AAH27432.1 | |||
| BC049238.1 | AAH49238.1 | |||
| BC052403.1 | AAH52403.1 |
Target Gene Details
Entrez Gene ID: | 98732 |
Gene Name: | RAB3 GTPase activating protein subunit 2 |
Gene Aliases: |
1110059F07Rik, 2010002H18Rik, 5830469C09, AI851069, AW743433, RAB3-GAP150, mKIAA0839 |
Location: |
Chr.1:185204088-185286746 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab3gap2 | NM_001163754.1 | NP_001157226.1 | ||
| AK004309.1 | ||||
| BQ176759.1 |
More Information
Set Membership: |
|

Genomic Map