Assay Details
Target Gene Details
Entrez Gene ID: | 214742 |
Gene Name: | REST corepressor 3 |
Gene Aliases: |
4921514E24Rik, C730034D20Rik, E130101E15Rik |
Location: |
Chr.1:192098546-192138067 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcor3 | NM_001290278.1 | 11 | 1566 | NP_001277207.1 |
| NM_001290282.1 | 11 | 1622 | NP_001277211.1 | |
| NM_144814.3 | 10 | 1508 | NP_659063.3 | |
| AK050287.1 | 11 | 1613 | ||
| AK135612.1 | 11 | 1539 | ||
| AK155440.1 | 11 | 1538 | ||
| BC057141.1 | 10 | 1255 | ||
| BC138938.1 | 10 | 1228 | ||
| BC138939.1 | 10 | 1228 |

Genomic Map