Assay Details
Target Gene Details
Entrez Gene ID: | 68002 |
Gene Name: | succinate dehydrogenase complex assembly factor 4 |
Gene Aliases: |
1110058L19Rik, 1700001E18Rik, C6orf57 |
Location: |
Chr.1:23995939-24005640 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdhaf4 | NM_026503.3 | 3 | 471 | NP_080779.2 |
| AK003789.1 | 3 | 429 | BAC25055.1 | |
| AK004301.1 | 3 | 451 | BAB23255.1 | |
| AK007909.1 | 3 | 464 | BAB25342.1 | |
| AK015824.1 | 3 | 472 | BAB29991.1 | |
| BC028981.1 | 3 | 436 | AAH28981.1 | |
| BF147474.1 | 1 | 205 |
More Information
Set Membership: |
|

Genomic Map