Assay Details
Target Gene Details
Entrez Gene ID: | 59125 |
Gene Name: | NIMA (never in mitosis gene a)-related expressed kinase 7 |
Gene Aliases: |
2810460C19Rik, AU020186 |
Location: |
Chr.1:138484106-138619779 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nek7 | NM_021605.3 | 10 | 2179 | NP_067618.1 |
| AF217650.1 | 9 | 2030 | AAG16652.1 | |
| AK035502.1 | 10 | 2178 | BAC29080.1 | |
| AK088173.1 | 10 | 2228 | BAC40190.1 | |
| AK165583.1 | 8 | 1948 | BAE38274.1 | |
| AK169649.1 | 1 | 1015 |
More Information
Set Membership: |
|

Genomic Map