Assay Details
Target Gene Details
Entrez Gene ID: | 23956 |
Gene Name: | neuraminidase 2 |
Gene Aliases: |
MBS, MSS, MTS |
Location: |
Chr.1:87574027-87597845 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neu2 | NM_001160163.1 | 4 | 853 | NP_001153635.1 |
| NM_001160164.1 | 3 | 751 | NP_001153636.1 | |
| NM_001160165.1 | 3 | 879 | NP_001153637.1 | |
| NM_015750.3 | 3 | 719 | NP_056565.1 | |
| AB028023.1 | 3 | 721 | BAA92867.1 | |
| AB048604.1 | 3 | 976 | BAB39152.1 | |
| AF139059.1 | 3 | 833 | ||
| AK009828.1 | 2 | 933 | ||
| AK145284.1 | 3 | 752 | BAE26342.1 | |
| BC026408.1 | 2 | 507 | AAH26408.1 | |
| BC105657.1 | 3 | 721 | ||
| BC105658.1 | 3 | 721 | ||
| BC105659.1 | 3 | 721 | ||
| BC105661.1 | 2 | 981 |
More Information
Set Membership: |
|

Genomic Map