Assay Details
Target Gene Details
Entrez Gene ID: | 241159 |
Gene Name: | sialidase 4 |
Gene Aliases: |
9330166I04 |
Location: |
Chr.1:94020446-94028334 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neu4 | NM_173772.3 | 3 | 489 | NP_776133.1 |
| AB510404.1 | 4 | 470 | ||
| AK034236.1 | 3 | 460 | BAC28641.1 | |
| AY258421.1 | 3 | 402 | AAP81169.1 | |
| BC070438.1 | 3 | 489 | AAH70438.1 |
More Information
Set Membership: |
|

Genomic Map