Assay Details
Target Gene Details
Entrez Gene ID: | 27280 |
Gene Name: | pleckstrin homology-like domain, family A, member 3 |
Gene Aliases: |
Tih1 |
Location: |
Chr.1:135766085-135769134 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phlda3 | NM_013750.2 | 1 | 672 | NP_038778.1 |
| AK003767.1 | 1 | 638 | BAB22985.1 | |
| AK187696.1 | ||||
| AK199905.1 | ||||
| BC023408.1 | 1 | 574 | AAH23408.1 |

Genomic Map