Assay Details
Target Gene Details
Entrez Gene ID: | 27280 |
Gene Name: | pleckstrin homology-like domain, family A, member 3 |
Gene Aliases: |
Tih1 |
Location: |
Chr.1:135766085-135769134 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phlda3 | NM_013750.2 | 2 | 1353 | NP_038778.1 |
| AI834921.1 | 1 | 131 | ||
| AK003767.1 | 2 | 1319 | BAB22985.1 | |
| AK189460.1 | 1 | 49 | ||
| AK195224.1 | 1 | 49 | ||
| AK218195.1 | 1 | 80 | ||
| BC023408.1 | 2 | 1255 | AAH23408.1 |

Genomic Map