Assay Details
Target Gene Details
Entrez Gene ID: | 67886 |
Gene Name: | calmodulin regulated spectrin-associated protein family, member 2 |
Gene Aliases: |
1600013L13Rik, 4930541M15Rik, Camsap1l1, mKIAA1078 |
Location: |
Chr.1:136268123-136346131 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camsap2 | NM_001081360.1 | 1 | 403 | NP_001074829.1 |
| AK016014.1 | 1 | 393 | ||
| AK028542.1 | 1 | 404 | BAC25999.2 | |
| AK053092.1 | 1 | 404 | ||
| BC138367.1 | ||||
| BC145258.1 |

Genomic Map