Assay Details
Target Gene Details
Entrez Gene ID: | 12477 |
Gene Name: | cytotoxic T-lymphocyte-associated protein 4 |
Gene Aliases: |
Cd152, Ctla-4, Ly-56 |
Location: |
Chr.1:60909025-60915832 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctla4 | NM_001281976.1 | 2 | 306 | NP_001268905.1 |
| NM_009843.4 | 2 | 306 | NP_033973.2 | |
| AB097214.1 | 2 | 160 | BAC41333.1 | |
| AK138891.1 | 2 | 239 | ||
| BC042741.1 | 2 | 306 | AAH42741.1 | |
| BC052683.1 | 2 | 226 | AAH52683.1 | |
| X05719.1 | 2 | 217 | CAA29191.1 |
More Information
Set Membership: |
|

Genomic Map