Assay Details
Target Gene Details
Entrez Gene ID: | 72750 |
Gene Name: | family with sequence similarity 117, member B |
Gene Aliases: |
2810425F24Rik, 6330416D14Rik, Als2cr13 |
Location: |
Chr.1:59913006-59985348 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam117b | NM_001037725.3 | 1 | 123 | NP_001032814.1 |
| AK154812.1 | 1 | 125 | BAE32846.1 |

Genomic Map