Assay Details
Target Gene Details
Entrez Gene ID: | 67099 |
Gene Name: | methyltransferase like 21A |
Gene Aliases: |
2310038H17Rik, AI464204, Fam119a |
Location: |
Chr.1:64606473-64620421 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl21a | NM_025964.3 | 1 | 151 | NP_080240.1 |
| AK009673.1 | BAB26430.1 | |||
| AK009675.1 | BAB26431.1 | |||
| AK009678.1 | BAB26434.1 | |||
| AK009679.1 | BAB26435.1 | |||
| AK009680.1 | BAB26436.1 | |||
| AK009682.1 | BAB26437.1 | |||
| AK011433.1 | BAB27618.1 | |||
| AK033578.1 | 1 | 152 | BAC28371.1 |

Genomic Map