Assay Details
Target Gene Details
Entrez Gene ID: | 67099 |
Gene Name: | methyltransferase like 21A |
Gene Aliases: |
2310038H17Rik, AI464204, Fam119a |
Location: |
Chr.1:64606473-64620421 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl21a | NM_025964.3 | 3 | 1562 | NP_080240.1 |
| AK009673.1 | 4 | 882 | BAB26430.1 | |
| AK009675.1 | 4 | 884 | BAB26431.1 | |
| AK009678.1 | 4 | 884 | BAB26434.1 | |
| AK009679.1 | 4 | 884 | BAB26435.1 | |
| AK009680.1 | 4 | 884 | BAB26436.1 | |
| AK009682.1 | 4 | 884 | BAB26437.1 | |
| AK011433.1 | 4 | 908 | BAB27618.1 | |
| AK033578.1 | 3 | 1563 | BAC28371.1 | |
| BC026952.1 | 3 | 832 | AAH26952.1 |

Genomic Map