Assay Details
Target Gene Details
Entrez Gene ID: | 241128 |
Gene Name: | family with sequence similarity 124, member B |
Gene Aliases: |
A830043J08Rik |
Location: |
Chr.1:80186969-80218504 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam124b | NM_173425.3 | 1 | 343 | NP_775601.1 |
| AK043867.1 | 1 | 344 | BAC31685.1 | |
| BC150872.1 | 1 | 228 |

Genomic Map