Assay Details
Target Gene Details
Entrez Gene ID: | 74013 |
Gene Name: | raftlin family member 2 |
Gene Aliases: |
2700010E02Rik, 3222401M22Rik |
Location: |
Chr.1:55170159-55227065 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rftn2 | NM_028713.1 | 9 | 2390 | NP_082989.1 |
| AK014318.1 | 9 | 2142 | BAB29269.1 | |
| BC038341.1 | 9 | 2390 | AAH38341.1 |

Genomic Map