Assay Details
Target Gene Details
Entrez Gene ID: | 140559 |
Gene Name: | immunoglobulin superfamily, member 8 |
Gene Aliases: |
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384 |
Location: |
Chr.1:172312407-172319841 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf8 | NM_080419.1 | 2 | 433 | NP_536344.1 |
| AF411055.1 | 2 | 304 | AAL02217.1 | |
| AF439263.1 | 2 | 416 | AAN63626.1 | |
| AK140254.1 | 2 | 446 | ||
| AK146104.1 | 2 | 446 | BAE26904.1 | |
| AK146221.1 | 2 | 452 | BAE26989.1 | |
| AK150239.1 | 2 | 452 | BAE29403.1 | |
| AK158522.1 | 2 | 454 | BAE34544.1 | |
| AK168363.1 | 2 | 452 | BAE40298.1 | |
| AK183814.1 | 1 | 90 | ||
| BC026464.1 | 1 | 239 | AAH26464.1 | |
| BC048387.1 | 2 | 433 | AAH48387.1 |
More Information
Set Membership: |
|

Genomic Map