Assay Details
Target Gene Details
Entrez Gene ID: | 381337 |
Gene Name: | family with sequence similarity 178, member B |
Gene Aliases: |
1700024G10Rik, Gm990 |
Location: |
Chr.1:36562696-36683190 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam178b | NM_001126046.1 | 15 | 2013 | NP_001119518.1 |
| NM_027957.1 | 6 | 636 | NP_082233.1 | |
| AK006300.1 | 6 | 637 | BAB24515.1 | |
| BC114576.1 | AAI14577.1 | |||
| BU605266.1 | 3 | 262 | ||
| BY445320.1 | 2 | 150 |

Genomic Map