Assay Details
Target Gene Details
Entrez Gene ID: | 16171 |
Gene Name: | interleukin 17A |
Gene Aliases: |
Ctla-8, Ctla8, IL-17, IL-17A, Il17 |
Location: |
Chr.1:20730905-20734496 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il17a | NM_010552.3 | 3 | 445 | NP_034682.1 |
| AK040420.1 | 3 | 446 | BAC30590.1 | |
| BC119303.1 | 3 | 445 | ||
| BC119309.1 | 3 | 445 | ||
| U43088.1 | 3 | 432 | AAB05222.1 |

Genomic Map