Assay Details
Target Gene Details
Entrez Gene ID: | 69962 |
Gene Name: | methyltransferase like 18 |
Gene Aliases: |
2810422O20Rik, C78571 |
Location: |
Chr.1:163994891-163997243 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl18 | NM_027279.2 | 2 | 783 | NP_081555.2 |
| AK013137.1 | 2 | 725 | BAB28672.1 | |
| AK139786.1 | 2 | 841 | BAE24137.1 | |
| BC050143.1 | 2 | 783 | AAH50143.1 | |
| BC052693.1 | 2 | 815 | AAH52693.1 |

Genomic Map