Assay Details
Target Gene Details
Entrez Gene ID: | 13244 |
Gene Name: | degenerative spermatocyte homolog 1 (Drosophila) |
Gene Aliases: |
AA536663, Degs, Des1, Mdes |
Location: |
Chr.1:182275770-182282759 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Degs1 | NM_007853.4 | 3 | 1640 | NP_031879.1 |
| AF466376.1 | 3 | 1657 | AAM12532.1 | |
| AK002617.1 | 3 | 1648 | BAB22233.1 | |
| AK077257.1 | 3 | 1641 | BAC36713.1 | |
| AK149811.1 | 3 | 1630 | BAE29098.1 | |
| BC003751.1 | 3 | 1658 | AAH03751.1 | |
| Y08460.1 | 3 | 1625 | CAA69714.1 |
More Information
Set Membership: |
|

Genomic Map