Assay Details
Target Gene Details
Entrez Gene ID: | 240899 |
Gene Name: | leucine rich repeat containing 52 |
Gene Aliases: |
4930413P14Rik |
Location: |
Chr.1:167445675-167466780 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc52 | NM_001013382.2 | 2 | 989 | NP_001013400.1 |
| BC087947.1 | 2 | 989 | AAH87947.1 |

Genomic Map