Assay Details
Target Gene Details
Entrez Gene ID: | 27993 |
Gene Name: | IMP4, U3 small nucleolar ribonucleoprotein, homolog (yeast) |
Gene Aliases: |
AA409888, AV031295, D1Wsu40e |
Location: |
Chr.1:34439829-34445747 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Imp4 | NM_178601.3 | 9 | 1319 | NP_848716.2 |
| AK004135.1 | 8 | 1221 | ||
| AK008178.1 | 9 | 1281 | ||
| AK010504.1 | 9 | 1280 | BAB26989.1 | |
| AK013334.1 | 9 | 1248 | BAB28797.1 | |
| AK018039.1 | 9 | 1276 | ||
| AK084933.1 | 9 | 1255 | BAC39313.1 | |
| AK151176.1 | 9 | 1298 | BAE30178.1 | |
| AK152214.1 | 9 | 1298 | BAE31042.1 | |
| AK166903.1 | 9 | 1269 | BAE39105.1 | |
| BC037602.1 | 8 | 1240 | AAH37602.1 | |
| BC069930.1 | 8 | 1229 | AAH69930.1 |
More Information
Set Membership: |
|

Genomic Map