Assay Details
Target Gene Details
Entrez Gene ID: | 16324 |
Gene Name: | inhibin beta-B |
Gene Aliases: |
- |
Location: |
Chr.1:119415465-119422248 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inhbb | NM_008381.3 | 2 | 2069 | NP_032407.1 |
| AK132352.1 | 2 | 2071 | BAE21120.1 | |
| BC048845.1 | 2 | 775 | AAH48845.1 | |
| X69620.1 | 1 | 40 | CAA49326.1 | |
| X83376.1 | 1 | 440 | CAA58290.1 |
More Information
Set Membership: |
|

Genomic Map