Assay Details
Target Gene Details
Entrez Gene ID: | 381306 |
Gene Name: | cDNA sequence BC055324 |
Gene Aliases: |
- |
Location: |
Chr.1:163945840-163994900 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC055324 | NM_201364.1 | 24 | 2665 | NP_958752.1 |
| AK031594.1 | 24 | 2661 | ||
| AK141661.1 | 24 | 2669 | BAE24790.1 | |
| BC052855.1 | 24 | 2659 | AAH52855.1 | |
| BC055324.1 | 24 | 2665 | AAH55324.1 |
Target Gene Details
Entrez Gene ID: | 240880 |
Gene Name: | SCY1-like 3 (S. cerevisiae) |
Gene Aliases: |
1200016D23Rik, 6030457O16, AW214499, Pace1 |
Location: |
Chr.1:163927958-163955126 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scyl3 | NM_001286002.1 | 14 | 3269 | NP_001272931.1 |
| NM_001286003.1 | 14 | 3520 | NP_001272932.1 | |
| NM_028776.5 | 14 | 3495 | NP_083052.1 | |
| AK030824.1 | 14 | 3470 | ||
| AK044100.1 | 14 | 3481 | BAC31777.1 | |
| AK050999.1 | 2 | 1030 | BAC34492.1 | |
| AK162937.1 | 14 | 3270 | BAE37123.1 | |
| BC043085.1 | 14 | 3196 | AAH43085.1 |
More Information
Set Membership: |
|

Genomic Map