Assay Details
Target Gene Details
Entrez Gene ID: | 78255 |
Gene Name: | Ral GEF with PH domain and SH3 binding motif 2 |
Gene Aliases: |
1810020P17Rik, 2210408F11Rik, 4921528G01Rik, 9130014M22Rik, AU043409, AW046161 |
Location: |
Chr.1:156804166-156941508 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgps2 | NM_001159965.1 | NP_001153437.1 | ||
| NM_001159966.1 | NP_001153438.1 | |||
| NM_001159967.1 | NP_001153439.1 | |||
| NM_001159968.1 | NP_001153440.1 | |||
| NM_023884.4 | NP_076373.3 | |||
| NR_027640.1 | ||||
| AK018622.1 | BAB31312.1 | |||
| AK019543.1 | ||||
| AK029049.1 | BAC26264.1 | |||
| AK029993.1 | ||||
| AK033549.1 | BAC28351.1 | |||
| AK036803.1 | BAC29580.1 | |||
| AK161080.1 | ||||
| AK169368.1 | BAE41116.1 | |||
| BC043132.1 | AAH43132.1 | |||
| BC052663.1 | AAH52663.1 | |||
| CJ104104.1 |
More Information
Set Membership: |
|

Genomic Map