Assay Details
Target Gene Details
Entrez Gene ID: | 109019 |
Gene Name: | nucleic acid binding protein 1 |
Gene Aliases: |
4930434H03Rik, 4930442A21Rik, 4930488J04Rik, 4933440J18Rik, 5830411E10Rik, AI852561, Nbp1, Obfc2a, Ssb2 |
Location: |
Chr.1:51465297-51478458 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nabp1 | NM_028696.3 | 1 | 481 | NP_082972.2 |
| AK015312.1 | BAE43243.1 | |||
| AK028886.1 | 1 | 482 | BAC26173.1 | |
| AK028936.1 | 1 | 149 | BAC26201.1 | |
| AK031015.1 | 1 | 483 | ||
| AK042351.1 | 1 | 482 | ||
| AK049396.1 | 1 | 482 | BAC33732.1 | |
| AK051607.1 | 1 | 482 | ||
| AK077226.1 | ||||
| AK077767.1 | 1 | 486 | ||
| AK079434.1 | 1 | 482 | ||
| AK083915.1 | 1 | 482 | BAC39059.1 | |
| AK163613.1 | 1 | 482 | BAE37421.1 | |
| AK166300.1 | 1 | 482 | BAE38689.1 | |
| AK167541.1 | BAE39608.1 | |||
| AK167652.1 | 1 | 482 | BAE39703.1 | |
| AY880264.1 | 1 | 482 | AAX68444.1 | |
| BC095967.1 | 1 | 147 | AAH95967.1 | |
| BC131923.1 | ||||
| BC132532.1 |
More Information
Set Membership: |
|

Genomic Map