Assay Details
Target Gene Details
Entrez Gene ID: | 19130 |
Gene Name: | prospero homeobox 1 |
Gene Aliases: |
A230003G05Rik |
Location: |
Chr.1:190118035-190170825 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prox1 | NM_008937.2 | 2 | 1913 | NP_032963.1 |
| AF061576.1 | 1 | 1500 | AAC32824.1 | |
| AK052889.1 | 2 | 1947 | BAC35190.1 | |
| AK143179.1 | 2 | 1737 | BAE25293.1 | |
| BC051411.1 | 2 | 1913 | AAH51411.1 |
More Information
Set Membership: |
|

Genomic Map