Assay Details
Target Gene Details
Entrez Gene ID: | 19225 |
Gene Name: | prostaglandin-endoperoxide synthase 2 |
Gene Aliases: |
COX2, Cox-2, PGHS-2, PHS-2, Pghs2, TIS10 |
Location: |
Chr.1:150100124-150108017 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgs2 | NM_011198.3 | 10 | 2830 | NP_035328.2 |
| AK144726.1 | 10 | 2855 | BAE26032.1 | |
| AK144956.1 | 10 | 2853 | BAE26154.1 | |
| AK149820.1 | 10 | 2855 | ||
| AK151293.1 | 1 | 351 | ||
| AK172161.1 | 10 | 2855 | BAE42855.1 | |
| M64291.1 | 10 | 2854 | AAA39924.1 | |
| M88242.1 | 10 | 2830 | AAA37740.1 | |
| X98792.1 | 1 | 772 |
More Information
Set Membership: |
|

Genomic Map