Assay Details
Target Gene Details
Entrez Gene ID: | 65247 |
Gene Name: | ankyrin repeat and SOCS box-containing 1 |
Gene Aliases: |
1700029O08Rik, 1700054C17Rik, AI426616, AI851378, mKIAA1146 |
Location: |
Chr.1:91539459-91559590 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb1 | NM_001039126.2 | 3 | 642 | NP_001034215.1 |
| NM_023046.4 | 3 | 443 | NP_075533.3 | |
| AF155352.1 | 3 | 394 | AAD38808.1 | |
| AK006515.1 | 3 | 444 | BAB24628.1 | |
| AK122456.1 | 3 | 498 | BAC65738.1 | |
| AK153774.1 | 3 | 664 | BAE32180.1 | |
| BC034869.1 | 4 | 437 | ||
| BU938650.1 | 3 | 446 | ||
| KC789642.1 | ||||
| KC789643.1 |

Genomic Map