Assay Details
Target Gene Details
Entrez Gene ID: | 65247 |
Gene Name: | ankyrin repeat and SOCS box-containing 1 |
Gene Aliases: |
1700029O08Rik, 1700054C17Rik, AI426616, AI851378, mKIAA1146 |
Location: |
Chr.1:91539459-91559590 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb1 | NM_001039126.2 | 5 | 2748 | NP_001034215.1 |
| NM_023046.4 | 5 | 2549 | NP_075533.3 | |
| AK080428.1 | 1 | 880 | ||
| AK122456.1 | 5 | 2604 | BAC65738.1 |

Genomic Map