Assay Details
Target Gene Details
Entrez Gene ID: | 14200 |
Gene Name: | four and a half LIM domains 2 |
Gene Aliases: |
FHL-2, SLIM-3, SLIM3 |
Location: |
Chr.1:43123071-43198299 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhl2 | NM_001289533.1 | 6 | 1126 | NP_001276462.1 |
| NM_010212.4 | 6 | 1275 | NP_034342.1 | |
| AF055889.1 | 5 | 1003 | AAC12770.1 | |
| AF153340.1 | 5 | 1047 | AAD34170.1 | |
| AK052936.1 | 6 | 1127 | BAC35211.1 | |
| AK133708.1 | 6 | 1247 | ||
| AK147024.1 | 6 | 1276 | BAE27617.1 | |
| BC021468.1 | 6 | 1184 | AAH21468.1 | |
| U77040.2 | 6 | 1165 | AAB19211.2 |
More Information
Set Membership: |
|

Genomic Map