Assay Details
Target Gene Details
Entrez Gene ID: | 66447 |
Gene Name: | microsomal glutathione S-transferase 3 |
Gene Aliases: |
2010012L10Rik, 2010306B17Rik, 2700004G04Rik, AA516734, GST-III |
Location: |
Chr.1:167372384-167393797 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgst3 | NM_025569.1 | 6 | 506 | NP_079845.1 |
| AK003246.1 | 6 | 506 | BAB22665.1 | |
| AK003309.1 | 6 | 492 | BAB22706.1 | |
| AK003476.1 | 6 | 499 | BAB22808.1 | |
| AK003492.1 | 6 | 499 | BAB22819.1 | |
| AK008211.1 | 5 | 496 | BAB25535.1 | |
| AK008533.1 | 6 | 495 | BAB25726.1 | |
| AK135742.1 | 6 | 481 | BAE22638.1 | |
| BC029669.1 | 5 | 485 | AAH29669.1 |
More Information
Set Membership: |
|

Genomic Map