Assay Details
Target Gene Details
Entrez Gene ID: | 258274 |
Gene Name: | olfactory receptor 1412 |
Gene Aliases: |
MOR208-4 |
Location: |
Chr.1:92588332-92589297 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr1412 | NM_146277.1 | 1 | 635 | NP_666389.1 |
| BC125241.1 | 1 | 635 |
More Information
Set Membership: |
|

Genomic Map