Assay Details
Target Gene Details
Entrez Gene ID: | 259039 |
Gene Name: | olfactory receptor 1413 |
Gene Aliases: |
MOR208-1 |
Location: |
Chr.1:92573125-92574206 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr1413 | NM_147037.2 | 1 | 369 | NP_667248.1 |
| BC116898.1 | 1 | 369 |
More Information
Set Membership: |
|

Genomic Map