Assay Details
Target Gene Details
Entrez Gene ID: | 71869 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 12 |
Gene Aliases: |
2300003F07Rik, 4833409F13Rik |
Location: |
Chr.1:106934449-106957080 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb12 | NM_001199213.1 | 8 | 1501 | NP_001186142.1 |
| NM_027971.2 | 8 | 1571 | NP_082247.1 | |
| AK009018.1 | 8 | 1572 | BAB26028.1 | |
| AK040697.1 | 8 | 1505 | BAC30672.1 | |
| BC062134.1 | 8 | 1476 | AAH62134.1 |
More Information
Set Membership: |
|

Genomic Map